missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Crotonate de méthyle 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Incolore |
| Boiling Point | 120°C |
| Quantity | 25 mL |
| UN Number | 3272 |
| Formula Weight | 100.12 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | Methyl Crotonate |
Sécurité et manipulation
| EINECSNumber | (2)-0966 |
| RTECSNumber | GQ5710000 |
| TSCA | Yes |