missing translation for 'onlineSavingsMsg'
Learn More

Methyl Carbazate 98.0+%, TCI America™

Catalog No. C1098500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1098500G 500 g
C109825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1098500G Supplier TCI America Supplier No. C1098500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6294-89-9
Molecular Formula C2H6N2O2
Molecular Weight (g/mol) 90.08
MDL Number MFCD00007594
InChI Key WFJRIDQGVSJLLH-UHFFFAOYSA-N
Synonym methyl carbazate, methyl hydrazinecarboxylate, methyl hydrazinocarboxylate, methoxycarbohydrazide, methylcarbazate, hydrazinecarboxylic acid, methyl ester, carbomethoxyhydrazide, methyl hydrazinoformate, methoxycarbonyl hydrazine, carbazic acid methyl ester
PubChem CID 80519
IUPAC Name methoxycarbohydrazide
SMILES COC(=O)NN

Specifications

Melting Point 73°C
Color White-Yellow
Boiling Point 108°C
Quantity 500 g
UN Number 2811
Formula Weight 90.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Carbazate