missing translation for 'onlineSavingsMsg'
Learn More

Methyl Carbamate 98.0+%, TCI America™

Catalog No. C1012500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1012500G 500 g
C101225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1012500G Supplier TCI America Supplier No. C1012500G

Chemical Identifiers

CAS 598-55-0
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00007964
InChI Key GTCAXTIRRLKXRU-UHFFFAOYSA-N
Synonym urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech
PubChem CID 11722
ChEBI CHEBI:76606
IUPAC Name methyl carbamate
SMILES COC(N)=O

Specifications

Melting Point 54°C
Boiling Point 177°C
Color White
Quantity 500 g
Formula Weight 75.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Carbamate
RTECSNumber FC2450000
TSCA Yes