missing translation for 'onlineSavingsMsg'
Learn More

Methyl carbamate, 99%

Catalog No. AAA1652336
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1652336 500 g
AAA1652318 50 g
AAA1652322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1652336 Supplier Thermo Scientific Chemicals Supplier No. A1652336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 598-55-0 | C2H5NO2 | 75.07 g/mol

Methyl carbamate was used in the synthesis of protected aminocyclopropanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl carbamate was used in the synthesis of protected aminocyclopropanes.

Solubility
Soluble in water(20g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, metals, alkali metals.

Chemical Identifiers

CAS 598-55-0
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00007964
InChI Key GTCAXTIRRLKXRU-UHFFFAOYSA-N
Synonym urethylane, carbamic acid, methyl ester, methylurethan, carbamic acid methyl ester, methylkarbamat, methylkarbamat czech, unii-9wfx634x2t, ccris 885, methylester kyseliny karbaminove, methylester kyseliny karbaminove czech
PubChem CID 11722
ChEBI CHEBI:76606
IUPAC Name methyl carbamate
SMILES COC(N)=O

Specifications

Melting Point 54°C to 56°C
Boiling Point 176°C to 178°C
Beilstein 635779
Merck Index 14,6036
Solubility Information Soluble in water(20g/L).
Density 1.14
Quantity 500 g
Flash Point >100°C (212°F)
Formula Weight 75.07
Percent Purity 99%
Chemical Name or Material Methyl carbamate
Show More Show Less
Hazard Category H315-H319-H335-H351
Hazard Statement GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P332+P313-P362-P501c
EINECSNumber 209-939-2
RTECSNumber FC2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only