missing translation for 'onlineSavingsMsg'
Learn More

Methyl butyrate, 99%

CAS: 623-42-7 | C5H10O2 | 102.13 g/mol

Supplier:  Thermo Scientific Chemicals 414141000

Quantity 100 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC414141000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

623-42-7
102.13
UUIQMZJEGPQKFD-UHFFFAOYSA-N
12180
CCCC(=O)OC
C5H10O2
MFCD00009391
methyl butyrate, butanoic acid, methyl ester, methyl n-butyrate, butyric acid, methyl ester, methyl n-butanoate, butyric acid methyl ester, methyl-n-butyrate, n-butyric acid methyl ester, methyl butyrate natural, unii-cgx598508o
methyl butanoate

Specifications

Methyl butyrate
1mg KOH/g max.
Colorless to Yellow
102°C
Authentic
Glass bottle
1.3850 to 1.39
100 g
0.897
methyl butyrate, butanoic acid, methyl ester, methyl n-butyrate, butyric acid, methyl ester, methyl n-butanoate, butyric acid methyl ester, methyl-n-butyrate, n-butyric acid methyl ester, methyl butyrate natural, unii-cgx598508o
UUIQMZJEGPQKFD-UHFFFAOYSA-N
methyl butanoate
12180
99%
99%
623-42-7
0.8970g/mL
12°C
98.5% min. (GC)
C5H10O2
CH3(CH2)2COOCH3
MFCD00009391
15, 6109
Solubility in water: insoluble
CCCC(=O)OC
102.13
102.13
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

EINECSNumber : 210-792-1

RTECSNumber : ET5500000

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions