missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl α-bromophenylacetate, 97+%, Thermo Scientific™
Chemical Identifiers
| CAS | 3042-81-7 |
|---|---|
| Molecular Formula | C9H9BrO2 |
| Molecular Weight (g/mol) | 229.07 |
| MDL Number | MFCD00013535 |
| InChI Key | NHFBYYMNJUMVOT-UHFFFAOYSA-N |
| Synonym | methyl alpha-bromophenylacetate, methyl bromophenylacetate, methyl bromo phenyl acetate, methyl .alpha.-bromophenylacetate, methyl2-bromo-2-phenylacetate, methyl .alpha.-bromobenzeneacetate, benzeneacetic acid, .alpha.-bromo-, methyl ester, +/--methyl alpha-bromophenylacetate, alpha-bromophenylacetic acid methyl ester, methyl .alpha.-bromo-.alpha.-phenylacetate |
| PubChem CID | 137806 |
| IUPAC Name | methyl 2-bromo-2-phenylacetate |
| SMILES | COC(=O)C(C1=CC=CC=C1)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC379300050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC379300250
|
N/A | 25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C9H9BrO2 | |
| MFCD00013535 | |
| methyl alpha-bromophenylacetate, methyl bromophenylacetate, methyl bromo phenyl acetate, methyl .alpha.-bromophenylacetate, methyl2-bromo-2-phenylacetate, methyl .alpha.-bromobenzeneacetate, benzeneacetic acid, .alpha.-bromo-, methyl ester, +/--methyl alpha-bromophenylacetate, alpha-bromophenylacetic acid methyl ester, methyl .alpha.-bromo-.alpha.-phenylacetate | |
| methyl 2-bromo-2-phenylacetate |
Specifications
| 3042-81-7 | |
| 113.0°C (3.0 mmHg) | |
| 97% min. (GC) | |
| C9H9BrO2 | |
| MFCD00013535 | |
| methyl alpha-bromophenylacetate, methyl bromophenylacetate, methyl bromo phenyl acetate, methyl .alpha.-bromophenylacetate, methyl2-bromo-2-phenylacetate, methyl .alpha.-bromobenzeneacetate, benzeneacetic acid, .alpha.-bromo-, methyl ester, +/--methyl alpha-bromophenylacetate, alpha-bromophenylacetic acid methyl ester, methyl .alpha.-bromo-.alpha.-phenylacetate | |
| NHFBYYMNJUMVOT-UHFFFAOYSA-N | |
| methyl 2-bromo-2-phenylacetate | |
| 137806 | |
| 97+% |
| 1.4550g/mL | |
| 113°C | |
| Glass bottle | |
| C6H5CH(Br)CO2CH3 | |
| 1.455 | |
| Solubility in water: .. Other solubilities: soluble in most organic solvents | |
| COC(=O)C(C1=CC=CC=C1)Br | |
| 229.07 | |
| 229.07 | |
| Methyl α-bromophenylacetate |
Safety and Handling
GHS Signal Word: Danger
RUO – Research Use Only