missing translation for 'onlineSavingsMsg'
Learn More

Methyl Bromodifluoroacetate 97.0+%, TCI America™

Catalog No. M20205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20205G 5 g
M202025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20205G Supplier TCI America Supplier No. M20205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 683-98-7
Molecular Formula C3H3BrF2O2
Molecular Weight (g/mol) 188.96
MDL Number MFCD06248118
InChI Key IOBCLXSGWMFVQJ-UHFFFAOYSA-N
Synonym Bromodifluoroacetic Acid Methyl Ester
PubChem CID 2783339
IUPAC Name methyl 2-bromo-2,2-difluoroacetate
SMILES COC(=O)C(F)(F)Br

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 g
UN Number 2924
Formula Weight 188.96
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Bromodifluoroacetate