missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl blue

Catalog No. AAH3772114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3772114 25 g
AAH3772122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3772114 Supplier Thermo Scientific Chemicals Supplier No. H3772114
Only null left

CAS: 28983-56-4 | C37H26N3Na2O9S3 | 798.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28983-56-4
Molecular Formula C37H26N3Na2O9S3
Molecular Weight (g/mol) 798.79
MDL Number MFCD00058509
InChI Key TUHAIJABPUJAEY-UHFFFAOYSA-K
Synonym Acid blue 93; C.I. 42780
PubChem CID 76956083
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1NC1=CC=C(C=C1)C(C1=CC=C(NC2=CC=CC=C2S([O-])(=O)=O)C=C1)=C1C=CC(C=C1)=NC1=CC=CC=C1S([O-])(=O)=O

Specifications

IUPAC Name disodium 2-{[4-({4-[(2-sulfonatophenyl)amino]phenyl}({4-[(2-sulfonatophenyl)imino]cyclohexa-2,5-dien-1-ylidene})methyl)phenyl]amino}benzene-1-sulfonate
Formula Weight 799.8
Odor Odorless
Color Blue
Quantity 25 g
Physical Form Powder
Chemical Name or Material Methyl blue
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 249-352-9
RTECSNumber DB5044700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only