missing translation for 'onlineSavingsMsg'
Learn More

Methyl beta-D-ribofuranoside, 98%

Catalog No. AAJ6682806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6682806 5 g
AAJ6682814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6682806 Supplier Thermo Scientific Chemicals Supplier No. J6682806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7473-45-2 | C6H12O5 | 164.157 g/mol

D-Ribose was converted into methyl 5-O-benzyl-beta-D-ribofuranoside and this, on tin-mediated allylation, gave a mixture of the 2-O-allyl and 3-O-allyl derivatives which were separated by chromatography. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Ribose was converted into methyl 5-O-benzyl-beta-D-ribofuranoside and this, on tin-mediated allylation, gave a mixture of the 2-O-allyl and 3-O-allyl derivatives which were separated by chromatography. 

Solubility
Soluble in water

Notes
Store at 2-8°C. Incompatible with heat, oxidizing agents.

Chemical Identifiers

CAS 7473-45-2
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.157
MDL Number MFCD00047075
InChI Key NALRCAPFICWVAQ-KVTDHHQDSA-N
Synonym methyl beta-d-ribofuranoside, methyl b-d-ribofuranoside, methyl, a-d-ribofuranoside, 2r,3s,4r,5r-2-hydroxymethyl-5-methoxytetrahydrofuran-3,4-diol, 2r,3s,4r,5r-2-hydroxymethyl-5-methoxyoxolane-3,4-diol, methyl pentofuranoside #, methyl .beta.-d-ribofuranoside, 1-o-methyl-beta-d-ribofuranoside, pubchem10614, methyl-b-d-ribofuranoside
PubChem CID 81983
IUPAC Name (2R,3S,4R,5R)-2-(hydroxymethyl)-5-methoxyoxolane-3,4-diol
SMILES COC1C(C(C(O1)CO)O)O

Specifications

Melting Point 78°C
Color White
Quantity 5 g
Solubility Information Soluble in water
Optical Rotation −50° (c=2 in Water)
Formula Weight 164.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl beta-D-ribofuranoside
EINECSNumber 231-271-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only