missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzoate 99.0+%, TCI America™

Catalog No. B0074500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0074500ML 500 mL
B007425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0074500ML Supplier TCI America Supplier No. B0074500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93-58-3
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00008421
InChI Key QPJVMBTYPHYUOC-UHFFFAOYSA-N
Synonym methylbenzoate, benzoic acid, methyl ester, clorius, benzoic acid methyl ester, niobe oil, oil of niobe, methyl benzenecarboxylate, essence of niobe, oniobe oil, oxidate le
PubChem CID 7150
ChEBI CHEBI:72775
IUPAC Name methyl benzoate
SMILES COC(=O)C1=CC=CC=C1

Specifications

Melting Point -12°C
Color Colorless
Boiling Point 200°C
Quantity 500 mL
Formula Weight 136.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Benzoate
EINECSNumber (3)-1356
RTECSNumber DH3850000
TSCA Yes