missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-D-Mannopyranoside 98.0+%, TCI America™

Catalog No. M036825G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M036825G 25 g
M0368250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M036825G Supplier TCI America Supplier No. M036825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 617-04-9 | C7H14O6 | 194.18 g/mol

Chemical Identifiers

CAS 617-04-9
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063262
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
Synonym methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
PubChem CID 101798
ChEBI CHEBI:43943
IUPAC Name 2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES COC1OC(CO)C(O)C(O)C1O

Specifications

Melting Point 192°C
Color White
Quantity 25 g
Formula Weight 194.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl alpha-D-Mannopyranoside