missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-Bromophenylacetate 98.0+%, TCI America™

Catalog No. M20385G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20385G 5 g
M203825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20385G Supplier TCI America Supplier No. M20385G
Only null left

Chemical Identifiers

CAS 3042-81-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00013535
InChI Key NHFBYYMNJUMVOT-UHFFFAOYSA-N
Synonym methyl alpha-bromophenylacetate, methyl bromophenylacetate, methyl bromo phenyl acetate, methyl .alpha.-bromophenylacetate, methyl2-bromo-2-phenylacetate, methyl .alpha.-bromobenzeneacetate, benzeneacetic acid, .alpha.-bromo-, methyl ester, +/--methyl alpha-bromophenylacetate, alpha-bromophenylacetic acid methyl ester, methyl .alpha.-bromo-.alpha.-phenylacetate
PubChem CID 137806
IUPAC Name methyl 2-bromo-2-phenylacetate
SMILES COC(=O)C(C1=CC=CC=C1)Br

Specifications

Color Yellow
Boiling Point 172°C
Quantity 5 g
UN Number 3265
Formula Weight 229.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl alpha-Bromophenylacetate
TSCA No
Recommended Storage Refrigerator