missing translation for 'onlineSavingsMsg'
Learn More

Methyl Acetate 99.5+%, TCI America™

Catalog No. S03005ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03005ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03005ML Supplier TCI America Supplier No. S03005ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79-20-9
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00008711
InChI Key KXKVLQRXCPHEJC-UHFFFAOYSA-N
Synonym tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
PubChem CID 6584
ChEBI CHEBI:77700
IUPAC Name methyl acetate
SMILES CC(=O)OC

Specifications

Color Colorless
Boiling Point 57°C
Quantity 5 mL
UN Number 1231
Formula Weight 74.08
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Acetate [Standard Material for GC]
EINECSNumber (2)-0725
RTECSNumber AI9100000
TSCA Yes