missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 7-phenylheptanoate, 98%
CAS: 101689-18-3 | C14H20O2 | 220.31 g/mol
Chemical Identifiers
| CAS | 101689-18-3 |
|---|---|
| Molecular Formula | C14H20O2 |
| Molecular Weight (g/mol) | 220.31 |
| MDL Number | MFCD02093971 |
| InChI Key | FZSNATXOGRAOIL-UHFFFAOYSA-N |
| Synonym | acmc-20akft, benzeneheptanoic acid,methyl ester, 7-phenylheptanoic acid methyl ester |
| PubChem CID | 7010318 |
| IUPAC Name | methyl 7-phenylheptanoate |
| SMILES | COC(=O)CCCCCCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAL1790503
|
N/A | 1 g |
N/A
|
|
|||||
|
AAL1790506
|
N/A | 5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 101689-18-3 | |
| 220.31 | |
| FZSNATXOGRAOIL-UHFFFAOYSA-N | |
| 7010318 | |
| COC(=O)CCCCCCC1=CC=CC=C1 |
| C14H20O2 | |
| MFCD02093971 | |
| acmc-20akft, benzeneheptanoic acid,methyl ester, 7-phenylheptanoic acid methyl ester | |
| methyl 7-phenylheptanoate |
Specifications
| 101689-18-3 | |
| C14H20O2 | |
| MFCD02093971 | |
| acmc-20akft, benzeneheptanoic acid,methyl ester, 7-phenylheptanoic acid methyl ester | |
| COC(=O)CCCCCCC1=CC=CC=C1 | |
| 220.31 | |
| 220.31 | |
| Methyl 7-phenylheptanoate |
| ∼155°C (1.5mmHg) | |
| 1.4946 | |
| 2694776 | |
| FZSNATXOGRAOIL-UHFFFAOYSA-N | |
| methyl 7-phenylheptanoate | |
| 7010318 | |
| 98% |
Safety and Handling
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only