missing translation for 'onlineSavingsMsg'
Learn More

Methyl 7-phenylheptanoate, 98%

Catalog No. AAL1790503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1790503 1 g
AAL1790506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1790503 Supplier Thermo Scientific Chemicals Supplier No. L1790503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 101689-18-3 | C14H20O2 | 220.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101689-18-3
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.31
MDL Number MFCD02093971
InChI Key FZSNATXOGRAOIL-UHFFFAOYSA-N
Synonym acmc-20akft, benzeneheptanoic acid,methyl ester, 7-phenylheptanoic acid methyl ester
PubChem CID 7010318
IUPAC Name methyl 7-phenylheptanoate
SMILES COC(=O)CCCCCCC1=CC=CC=C1

Specifications

Boiling Point ∼155°C (1.5mmHg)
Refractive Index 1.4946
Quantity 1 g
Beilstein 2694776
Formula Weight 220.31
Percent Purity 98%
Chemical Name or Material Methyl 7-phenylheptanoate
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only