accessibility menu, dialog, popup

UserName

CAS RN 101689-18-3

IUPAC Name: methyl 7-phenylheptanoate
Synonyms: methyl 7-phenylheptanoate
Molecular Weight (g/mol): 220.31
Molecular Formula: C14H20O2
InChi Key: FZSNATXOGRAOIL-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)