missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Iodosalicylate 98.0+%, TCI America™

Catalog No. p-7131470
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05395G 5 g
I053925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05395G Supplier TCI America Supplier No. I05395G

Chemical Identifiers

CAS 4068-75-1
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD00016462
InChI Key NRSWJTRJHPRZMH-UHFFFAOYSA-N
Synonym methyl 5-iodosalicylate, 5-iodosalicylic acid methyl ester, 2-hydroxy-5-iodobenzoic acid methyl ester, benzoic acid, 2-hydroxy-5-iodo-, methyl ester, methyl 2-hydroxy-5-iodobenzenecarboxylate, salicylic acid, 5-iodo-, methyl ester, methyl5-iodosalicylate, methyl-5-iodosalicylate, acmc-209jg0, methyl2-hydroxy-5-iodobenzoate
PubChem CID 248910
IUPAC Name methyl 2-hydroxy-5-iodobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)I)O

Specifications

Melting Point 77°C
Color White
Quantity 5 g
Formula Weight 278.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Iodosalicylate