missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Hexenoate 98.0+%, TCI America™

Catalog No. H08725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08725G 5 g
H087225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08725G Supplier TCI America Supplier No. H08725G
Only null left

Chemical Identifiers

CAS 2396-80-7
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00671538
InChI Key ASKDFGVMJZMYEM-UHFFFAOYSA-N
Synonym 5-Hexenoic Acid Methyl Ester
PubChem CID 520082
IUPAC Name methyl hex-5-enoate
SMILES COC(=O)CCCC=C

Specifications

Color Colorless
Boiling Point 157°C
Quantity 5 g
UN Number 3272
Formula Weight 128.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 5-Hexenoate