missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromovalerate, 97%

CAS: 5454-83-1 | C6H11BrO2 | 195.06 g/mol

$92.63 - $896.59

Chemical Identifiers

CAS 5454-83-1
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00000265
InChI Key RAVVJKCSZXAIQP-UHFFFAOYSA-N
PubChem CID 79557
IUPAC Name methyl 5-bromopentanoate
SMILES COC(=O)CCCCBr
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1907206
View Documents Promotion Details
N/A 5 g
Each for $92.63
Only null left
 
AAA1907214
View Documents Promotion Details
N/A 25 g
Each for $320.34
Only null left
 
AAA1907222
View Documents Promotion Details
N/A 100 g
Each for $896.59
Only null left
 
Description

Description

It is employed as an intermediate in the production of Apixaban .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate in the production of Apixaban .

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

5454-83-1
195.06
RAVVJKCSZXAIQP-UHFFFAOYSA-N
methyl 5-bromopentanoate
C6H11BrO2
MFCD00000265
79557
COC(=O)CCCCBr

Specifications

5454-83-1
79°C to 80°C (4 mmHg)
C6H11BrO2
MFCD00000265
methyl 5-bromovalerate, 5-bromovaleric acid methyl ester, pentanoic acid, 5-bromo-, methyl ester, methyl .omega.-bromopentanoate, 5-bromopentanoic acid methyl ester, 5-bromo-pentanoic acid methyl ester, methyl omega-bromopentanoate, methyl5-bromovalerate, valeric acid, 5-bromo-, methyl ester, methyl-5-bromovalerate
RAVVJKCSZXAIQP-UHFFFAOYSA-N
methyl 5-bromopentanoate
79557
97%
1.363
99°C (210°F)
1.463
1749649
Slightly soluble in water.
COC(=O)CCCCBr
195.06
195.06
Methyl 5-bromovalerate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 226-709-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only