accessibility menu, dialog, popup

UserName

CAS RN 5454-83-1

IUPAC Name: methyl 5-bromopentanoate
Synonyms: methyl 5-bromopentanoate
Molecular Weight (g/mol): 195.06
Molecular Formula: C6H11BrO2
InChi Key: RAVVJKCSZXAIQP-UHFFFAOYSA-N
SMILES: COC(=O)CCCCBr
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)