missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromovalerate, 97%

Catalog No. AAA1907206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1907206 5 g
AAA1907214 25 g
AAA1907222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1907206 Supplier Thermo Scientific Chemicals Supplier No. A1907206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5454-83-1 | C6H11BrO2 | 195.06 g/mol

It is employed as an intermediate in the production of Apixaban .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate in the production of Apixaban .

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 5454-83-1
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00000265
InChI Key RAVVJKCSZXAIQP-UHFFFAOYSA-N
PubChem CID 79557
IUPAC Name methyl 5-bromopentanoate
SMILES COC(=O)CCCCBr

Specifications

Density 1.363
Boiling Point 79°C to 80°C (4 mmHg)
Flash Point 99°C (210°F)
Refractive Index 1.463
Quantity 5 g
Beilstein 1749649
Synonym methyl 5-bromovalerate, 5-bromovaleric acid methyl ester, pentanoic acid, 5-bromo-, methyl ester, methyl .omega.-bromopentanoate, 5-bromopentanoic acid methyl ester, 5-bromo-pentanoic acid methyl ester, methyl omega-bromopentanoate, methyl5-bromovalerate, valeric acid, 5-bromo-, methyl ester, methyl-5-bromovalerate
Solubility Information Slightly soluble in water.
Formula Weight 195.06
Percent Purity 97%
Chemical Name or Material Methyl 5-bromovalerate
Show More Show Less
EINECSNumber 226-709-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only