missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Methoxysalicylate 99.0+%, TCI America™

Catalog No. M17975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17975G 5 g
M179725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17975G Supplier TCI America Supplier No. M17975G

Chemical Identifiers

CAS 5446-02-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00008424
InChI Key ZICRWXFGZCVTBZ-UHFFFAOYSA-N
Synonym methyl 4-methoxysalicylate, 2-hydroxy-4-methoxy-benzoic acid methyl ester, methyl-4-methoxysalicylate, benzoic acid, 2-hydroxy-4-methoxy-, methyl ester, 2-hydroxy-4-methoxybenzoic acid methyl ester, 4-methoxysalicylic acid methyl ester, acmc-2097hp, methyl 2-hydroxy-p-anisate, ksc494a7p, 2-methoxycarbonyl-5-methoxyphenol
PubChem CID 79523
IUPAC Name methyl 2-hydroxy-4-methoxybenzoate
SMILES COC1=CC(=C(C=C1)C(=O)OC)O

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 134°C
Quantity 5 g
Formula Weight 182.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Methoxysalicylate