missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methoxypyridine-2-carboxylate, 98%

Catalog No. AAH6451914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6451914 25 g
AAH6451906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6451914 Supplier Thermo Scientific Chemicals Supplier No. H6451914

CAS: 29681-43-4 | C8H9NO3 | 167.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29681-43-4
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00956223
InChI Key OJDKENGKKYVJLY-UHFFFAOYSA-N
Synonym methyl 4-methoxypicolinate, 4-methoxy-pyridine-2-carboxylic acid methyl ester, 4-methoxy-pyridine-2-carboxylicacidmethylester, 2-pyridinecarboxylic acid, 4-methoxy-, methyl ester, 4-methoxypicolinic acid methyl ester, 4-methoxypyridine-2-carboxylic acid methyl ester, methyl 4-methoxy-2-pyridinecarboxylate, 4-methoxy-pyridine-2-carboxylic acid metyl ester, pubchem11083, acmc-1b7sb
PubChem CID 1519345
IUPAC Name methyl 4-methoxypyridine-2-carboxylate
SMILES COC1=CC(=NC=C1)C(=O)OC

Specifications

Melting Point 47°C to 50°C
Color White to Yellow
Quantity 25 g
Beilstein 473379
Formula Weight 167.16
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 4-methoxypyridine-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only