missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(hydroxymethyl)benzoate, 98%, Thermo Scientific™

Numéro de catalogue. AC330840100
Modifier l'affichage
Click to view available options
10g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
AC330840100 10g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC330840100 Fournisseur Thermo Scientific Chemicals Code fournisseur 330840100

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Nom ou substance chimique Methyl 4-(hydroxymethyl)benzoate
Nom Remarque 98%
CAS 6908-41-4
Point d’éclair >110°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 98%
Emballage Glass bottle
Formule moléculaire C9H10O3
Quantité 10g
Numéro MDL MFCD00009984
Synonyme methyl 4-hydroxymethyl benzoate, benzoic acid, 4-hydroxymethyl-, methyl ester, 4-carbomethoxybenzyl alcohol, 4-hydroxymethyl benzoic acid methyl ester, methyl p-hydroxymethyl benzoate, acmc-209o5w, methyl p-hydroxymethylbenzoate, methyl-4-hydroxymethylbenzoate, p-methoxycarbonylbenzyl alcohol
Renseignements sur la solubilité Solubility in water: slightly soluble in water
Clé InChI VBWFYEFYHJRJER-UHFFFAOYSA-N
SOURIRES COC(=O)C1=CC=C(C=C1)CO
Nom de l’IUPAC methyl 4-(hydroxymethyl)benzoate
Poids moléculaire (g/mol) 166.18
PubChem CID 81325
Poids de formule 166.18
Pourcentage de pureté 98%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-017-0
TSCA TSCA

RUO – Research Use Only