missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluoro-3-nitrobenzoate, 98%

Catalog No. AAH6154214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6154214 25 g
AAH6154222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6154214 Supplier Thermo Scientific Chemicals Supplier No. H6154214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 329-59-9 | C8H6FNO4 | 199.14 g/mol

Methyl 4-fluoro-3-nitrobenzoate is used to prepare dimethyl 3-nitro-3',4-oxydibenzoate by reacting with 3-hydroxy-benzoic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-fluoro-3-nitrobenzoate is used to prepare dimethyl 3-nitro-3′,4-oxydibenzoate by reacting with 3-hydroxy-benzoic acid methyl ester.

Solubility
Soluble in ethanol, ether and methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents and strong bases.

Chemical Identifiers

CAS 329-59-9
Molecular Formula C8H6FNO4
Molecular Weight (g/mol) 199.14
MDL Number MFCD00196161
InChI Key CNJJSTPBUHAEFH-UHFFFAOYSA-N
Synonym methyl 4-fluoro-3-nitro-benzoate, 4-fluoro-3-nitrobenzoic acid methyl ester, 4-fluoro-3-nitro-benzoic acid methyl ester, benzoic acid, 4-fluoro-3-nitro-, methyl ester, pubchem2045, acmc-209hxf, methyl 4-fluoro-3 nitrobenzoate, 4-carbomethoxy-2-nitrofluorobenzene, methyl4-fluoro-3-nitrobenzoate
PubChem CID 5219721
IUPAC Name methyl 4-fluoro-3-nitrobenzoate
SMILES COC(=O)C1=CC(=C(F)C=C1)[N+]([O-])=O

Specifications

Melting Point 56°C to 59°C
Boiling Point 109°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Solubility Information Soluble in ethanol,ether and methanol. Insoluble in water.
Formula Weight 199.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 4-fluoro-3-nitrobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only