missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-(Cyanomethyl)benzoate 95.0+%, TCI America™
$219.48 - $662.26
Chemical Identifiers
| CAS | 76469-88-0 |
|---|---|
| Molecular Formula | C10H9NO2 |
| Molecular Weight (g/mol) | 175.19 |
| MDL Number | MFCD00060695 |
| InChI Key | XRZGMNGGCZTNGE-UHFFFAOYSA-N |
| Synonym | 4-(Cyanomethyl)benzoic Acid Methyl Ester |
| PubChem CID | 848548 |
| IUPAC Name | methyl 4-(cyanomethyl)benzoate |
| SMILES | COC(=O)C1=CC=C(CC#N)C=C1 |
Chemical Identifiers
| 76469-88-0 | |
| 175.19 | |
| XRZGMNGGCZTNGE-UHFFFAOYSA-N | |
| 848548 | |
| COC(=O)C1=CC=C(CC#N)C=C1 |
| C10H9NO2 | |
| MFCD00060695 | |
| 4-(Cyanomethyl)benzoic Acid Methyl Ester | |
| methyl 4-(cyanomethyl)benzoate |
Specifications
| 76469-88-0 | |
| 112°C | |
| MFCD00060695 | |
| 4-(Cyanomethyl)benzoic Acid Methyl Ester | |
| COC(=O)C1=CC=C(CC#N)C=C1 | |
| 175.19 | |
| 175.19 | |
| Crystalline Powder |
| 62°C | |
| C10H9NO2 | |
| 5 g | |
| XRZGMNGGCZTNGE-UHFFFAOYSA-N | |
| methyl 4-(cyanomethyl)benzoate | |
| 848548 | |
| ≥95.0% (GC) | |
| Methyl 4-(Cyanomethyl)benzoate |
Safety and Handling
TSCA : No