missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(Cyanomethyl)benzoate 95.0+%, TCI America™

Catalog No. m23415g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
M23415G 5 g
M234125G 25 g
2 options

Catalog No. M23415G

Supplier: TCI America M23415G

Only null left

Chemical Identifiers

CAS 76469-88-0
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00060695
InChI Key XRZGMNGGCZTNGE-UHFFFAOYSA-N
Synonym 4-(Cyanomethyl)benzoic Acid Methyl Ester
PubChem CID 848548
IUPAC Name methyl 4-(cyanomethyl)benzoate
SMILES COC(=O)C1=CC=C(CC#N)C=C1

Specifications

Melting Point 62°C
Boiling Point 112°C
Quantity 5 g
Formula Weight 175.19
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-(Cyanomethyl)benzoate