missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(Cyanomethyl)benzoate 95.0+%, TCI America™

Catalog No. m234125g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M234125G 25 g
M23415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M234125G Supplier TCI America Supplier No. M234125G

Chemical Identifiers

CAS 76469-88-0
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00060695
InChI Key XRZGMNGGCZTNGE-UHFFFAOYSA-N
Synonym 4-(Cyanomethyl)benzoic Acid Methyl Ester
PubChem CID 848548
IUPAC Name methyl 4-(cyanomethyl)benzoate
SMILES COC(=O)C1=CC=C(CC#N)C=C1

Specifications

Melting Point 62°C
Boiling Point 112°C
Quantity 25 g
Formula Weight 175.19
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-(Cyanomethyl)benzoate