missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(bromomethyl)benzoate, 98%, Thermo Scientific™

Catalog No. AC306101000
Change view
Click to view available options
Glass bottle; 100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC306101000 Glass bottle; 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC306101000 Supplier Thermo Scientific Chemicals Supplier No. 306101000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 2417-72-3
Boiling Point 130.0°C to 135.0°C (2.0mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H9BrO2
Linear Formula BrCH2C6H4CO2CH3
Quantity 100g
MDL Number MFCD00032453
Synonym methyl 4-bromomethyl benzoate, 4-bromomethylbenzoic acid methyl ester, methyl4-bromomethyl benzoate, benzoic acid, 4-bromomethyl-, methyl ester, 4-bromomethyl benzoic acid methyl ester, p-methoxycarbonyl benzyl bromide, methyl p-bromomethyl benzoate, 4-bromomethyl-benzoic acid methyl ester, 4-carbomethoxybenzyl bromide
InChI Key NLWBJPPMPLPZIE-UHFFFAOYSA-N
SMILES COC(=O)C1=CC=C(CBr)C=C1
IUPAC Name methyl 4-(bromomethyl)benzoate
Molecular Weight (g/mol) 229.07
PubChem CID 256687
Formula Weight 229.07
Percent Purity 98%
Chemical Name or Material Methyl 4-(bromomethyl)benzoate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only