missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(bromomethyl)benzoate, 98%

Catalog No. AAA1055722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1055722 100 g
AAA1055706 5 g
AAA1055714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1055722 Supplier Thermo Scientific Chemicals Supplier No. A1055722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2417-72-3 | C9H9BrO2 | 229.07 g/mol

Methyl 4-(bromomethyl)benzoate is used in the preparation of potential anti-HIV agents. It i s also used as a catalyst for rearrangement of benzylthiothiazoline derivatives in the preparation aldose reductase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-(bromomethyl)benzoate is used in the preparation of potential anti-HIV agents. It i s also used as a catalyst for rearrangement of benzylthiothiazoline derivatives in the preparation aldose reductase inhibitors.

Solubility
Lowly soluble in water, highly soluble in ethanol and ether, not soluble in halogenated organic solvents

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Handle and store under inert gas. Keep away from strong oxidizing agents, strong bases. Stable under recommended storage conditions.

Chemical Identifiers

CAS 2417-72-3
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00032453
InChI Key NLWBJPPMPLPZIE-UHFFFAOYSA-N
PubChem CID 256687
IUPAC Name methyl 4-(bromomethyl)benzoate
SMILES COC(=O)C1=CC=C(CBr)C=C1

Specifications

Melting Point 53°C to 58°C
Boiling Point 130°C to 135°C (2 mmHg)
Quantity 100 g
UN Number UN1759
Beilstein 1867272
Synonym methyl 4-bromomethyl benzoate, 4-bromomethylbenzoic acid methyl ester, methyl4-bromomethyl benzoate, benzoic acid, 4-bromomethyl-, methyl ester, 4-bromomethyl benzoic acid methyl ester, p-methoxycarbonyl benzyl bromide, methyl p-bromomethyl benzoate, 4-bromomethyl-benzoic acid methyl ester, 4-carbomethoxybenzyl bromide
Solubility Information Lowly soluble in water,highly soluble in ethanol and ether,not soluble in halogenated organic solvents
Formula Weight 229.07
Percent Purity 98%
Chemical Name or Material Methyl 4- (bromomethyl)benzoate
Hazard Category H302-H314-H334-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P285-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 000-000-0
TSCA No

RUO – Research Use Only