missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Bromobenzoate 98.0+%, TCI America™

Catalog No. B055610G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B055610G 10 g
B055625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B055610G Supplier TCI America Supplier No. B055610G

Chemical Identifiers

CAS 619-42-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00013531
InChI Key CZNGTXVOZOWWKM-UHFFFAOYSA-N
Synonym benzoic acid, 4-bromo-, methyl ester, methyl p-bromobenzoate, 4-bromobenzoic acid methyl ester, p-bromobenzoic acid, methyl ester, unii-ub1ysj5xar, ub1ysj5xar, benzoic acid, p-bromo-, methyl ester, pubchem3962, methyl-4-bromobenzoate, methyl p-bromo benzoate
PubChem CID 12081
IUPAC Name methyl 4-bromobenzoate
SMILES COC(=O)C1=CC=C(C=C1)Br

Specifications

Melting Point 79°C
Color White
Quantity 10 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Bromobenzoate