missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,6-O-Benzylidene-alpha-D-glucopyranoside 98.0+%, TCI America™

Catalog No. M11255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11255G 5 g
M112525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11255G Supplier TCI America Supplier No. M11255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3162-96-7
Molecular Formula C14H18O6
Molecular Weight (g/mol) 282.292
MDL Number MFCD00006819
InChI Key VVSWDMJYIDBTMV-BTZLDLHRSA-N
Synonym 4ar,6s,7r,8r,8as-6-methoxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxine-7,8-diol, methyl 4,6-o-benzylidene-alpha-d-glucopyranoside, methyl 4,6-o-benzylidene-a-d-glucopyranoside, 4ar,6s,7r,8r,8as-6-methoxy-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxine-7,8-diol, methyl 4,6-o-benzylidenehexopyranoside #, zlchem 511, methyl 4,6-o-benzylidene-, a-d-glucopyranoside, methyl 4,6-o-phenylmethylene-, glucopyranoside, methyl 4,6-o-benzylidene-, .alpha.-d, .alpha.-d-glucopyranoside, methyl 4,6-o-phenylmethylene
PubChem CID 11822086
IUPAC Name (4aR,6S,7R,8R,8aS)-6-methoxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine-7,8-diol
SMILES COC1C(C(C2C(O1)COC(O2)C3=CC=CC=C3)O)O

Specifications

Melting Point 164°C
Color White-Yellow
Quantity 5 g
Formula Weight 282.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4,6-O-Benzylidene-alpha-D-glucopyranoside