missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, 97%

Catalog No. AAA1341822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1341822 100 g
AAA1341806 5 g
AAA1341814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1341822 Supplier Thermo Scientific Chemicals Supplier No. A1341822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3162-96-7 | C14H18O6 | 282.292 g/mol

Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, is used as a chiral building block and important intermediate in the preparation of different sugars. It is also used as an intermediate in chemical, pharmaceutical intermediate, pharmaceutical research and reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, is used as a chiral building block and important intermediate in the preparation of different sugars. It is also used as an intermediate in chemical, pharmaceutical intermediate, pharmaceutical research and reagent.

Solubility
It is soluble in water, acetone, benzene, ethyl ether

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 3162-96-7
Molecular Formula C14H18O6
Molecular Weight (g/mol) 282.292
MDL Number MFCD00006819
InChI Key VVSWDMJYIDBTMV-BTZLDLHRSA-N
Synonym 4ar,6s,7r,8r,8as-6-methoxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxine-7,8-diol, methyl 4,6-o-benzylidene-alpha-d-glucopyranoside, methyl 4,6-o-benzylidene-a-d-glucopyranoside, 4ar,6s,7r,8r,8as-6-methoxy-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxine-7,8-diol, methyl 4,6-o-benzylidenehexopyranoside #, zlchem 511, methyl 4,6-o-benzylidene-, a-d-glucopyranoside, methyl 4,6-o-phenylmethylene-, glucopyranoside, methyl 4,6-o-benzylidene-, .alpha.-d, .alpha.-d-glucopyranoside, methyl 4,6-o-phenylmethylene
PubChem CID 11822086
IUPAC Name (4aR,6S,7R,8R,8aS)-6-methoxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine-7,8-diol
SMILES COC1C(C(C2C(O1)COC(O2)C3=CC=CC=C3)O)O

Specifications

Melting Point 163°C to 166°C
Quantity 100 g
Beilstein 1291458
Sensitivity Hygroscopic
Solubility Information It is soluble in water,acetone,benzene,ethyl ether
Optical Rotation +110° (c=2 in Chloroform)
Formula Weight 282.29
Percent Purity 97%
Chemical Name or Material Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside
EINECSNumber 221-615-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only