missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,4,4-trifluoroacetoacetate, 95%

Catalog No. AAL1014014
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1014014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1014014 Supplier Thermo Scientific Chemicals Supplier No. L1014014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 83643-84-9 | C5H5F3O3 | 170.087 g/mol

It is used as a drug intermediate. It is a cyclization, oxidation and halogenating reaction catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a drug intermediate. It is a cyclization, oxidation and halogenating reaction catalyst.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.

Chemical Identifiers

CAS 83643-84-9
Molecular Formula C5H5F3O3
Molecular Weight (g/mol) 170.087
MDL Number MFCD00041004
InChI Key LKMUBWWZTSZGGV-UHFFFAOYSA-N
PubChem CID 550266
IUPAC Name methyl 4,4,4-trifluoro-3-oxobutanoate
SMILES COC(=O)CC(=O)C(F)(F)F

Specifications

Density 1.34
Boiling Point 117°C to 118°C
Flash Point 40°C (104°F)
Refractive Index 1.372
Quantity 25 g
UN Number UN3272
Beilstein 1777343
Synonym methyl trifluoroacetoacetate, methyl 4,4,4-trifluoroacetoacetate, 4,4,4-trifluoroacetoacetic acid methyl ester, methyl-4,4,4-trifluoro-3-oxobutanoate, butanoic acid, 4,4,4-trifluoro-3-oxo-, methyl ester, acmc-209psv, methyltri-fluoroacetoacetate, methyl4,4,4-trifluoroacetoacetate, methyl-4,4,4-trifluoroacetoacetate, methyl 3-oxo-4,4,4-trifluorobutyrate
Formula Weight 170.09
Percent Purity 95%
Chemical Name or Material Methyl 4,4,4-trifluoroacetoacetate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only