missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 98.0+%, TCI America™

Catalog No. M30571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30571G 1 g
M30575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30571G Supplier TCI America Supplier No. M30571G

Chemical Identifiers

CAS 171364-80-0
Molecular Formula C14H19BO4
Molecular Weight (g/mol) 262.11
MDL Number MFCD02179438
InChI Key REIZEQZILPXYKS-UHFFFAOYSA-N
Synonym methyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-methoxycarbonylphenylboronic acid pinacol ester, 4-methoxycarbonylphenylboronic acid pinacolate, 4-carbomethoxyphenylboronic acid pinacol ester, 4-methoxycarbonylphenylboronic acid, pinacol ester, 4-methoxycarbonyl phenylboronic acid pinacol ester, methyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzoic acid methyl ester, 4-methoxycarbonyl benzeneboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid methyl ester
PubChem CID 2773500
IUPAC Name methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 82°C
Color White
Quantity 1 g
Formula Weight 262.11
Percent Purity ≥98.0% (GC,T)
Physical Form Powder or Lumps
Chemical Name or Material Methyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate