missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Oxo-4-phenylbutyrate (mixture of isomers) 96.0+%, TCI America™

Catalog No. p-7133203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23961G 1 g
M23965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23961G Supplier TCI America Supplier No. M23961G

Chemical Identifiers

CAS 37779-49-0
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD03424746
InChI Key DTMSEOVTDVSPDO-UHFFFAOYSA-N
Synonym 3-Oxo-4-phenylbutyric Acid Methyl Ester, Methyl 4-Phenylacetoacetate, 4-Phenylacetoacetic Acid Methyl Ester
PubChem CID 2760261
IUPAC Name methyl 3-oxo-4-phenylbutanoate
SMILES COC(=O)CC(=O)CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 136°C
Quantity 1 g
Formula Weight 192.21
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Oxo-4-phenylbutyrate (mixture of isomers)