missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-nitrobenzoate, 98%

Catalog No. AC168445000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168445000 500 g Plastic bottle
AC168440250 25 g Plastic bottle
AC168441000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC168445000 Supplier Thermo Scientific Chemicals Supplier No. 168445000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 618-95-1 | C8H7NO4 | 181.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 618-95-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007250
InChI Key AXLYJLKKPUICKV-UHFFFAOYSA-N
Synonym 3-nitrobenzoic acid methyl ester, methyl m-nitrobenzoate, methyl3-nitrobenzoate, benzoic acid, 3-nitro-, methyl ester, m-nitrobenzoic acid, methyl ester, benzoic acid, m-nitro-, methyl ester, 3-nitro-benzoic acid methyl ester, 3-nitro-benzoicacimethylester, acmc-209mxc, methyl 3-nitro-benzoate
PubChem CID 69260
IUPAC Name methyl 3-nitrobenzoate
SMILES COC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 76°C to 80°C
Color Beige
Boiling Point 279°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula O2NC6H4CO2CH3
Quantity 500 g
Beilstein 09, 378
Merck Index 10, 6435
Solubility Information Solubility in water: insoluble
Formula Weight 181.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-nitrobenzoate
Show More Show Less
EINECSNumber 210-573-

RUO – Research Use Only