missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Methylthiophene-2-carboxylate 97.0+%, TCI America™

Catalog No. M26625G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26625G 5 g
M26621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26625G Supplier TCI America Supplier No. M26625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81452-54-2
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.20
MDL Number MFCD00234332
InChI Key BRWROFVPMUPMJQ-UHFFFAOYSA-N
Synonym methyl 3-methyl-2-thiophenecarboxylate, 3-methylthiophene-2-carboxylic acid methyl ester, 3-methyl-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-methyl-, methyl ester, methyl 3-methyl-thiophene-2-carboxylate, d methyl ester, 3-methyl-thiophene-2-carboxylic acid, pubchem10098, pubchem22915, ksc495k9b
PubChem CID 580757
IUPAC Name methyl 3-methylthiophene-2-carboxylate
SMILES COC(=O)C1=C(C)C=CS1

Specifications

Color Yellow
Boiling Point 217°C
Quantity 5 g
Formula Weight 156.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Methylthiophene-2-carboxylate