missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methyl-2-furoate, 98%

Catalog No. AAA1054114
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1054114 25 g
AAA1054106 5 g
AAA1054109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1054114 Supplier Thermo Scientific Chemicals Supplier No. A1054114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6141-57-7 | C7H8O3 | 140.14 g/mol

Methyl 3-methyl-2-furoate is an important raw material used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.It is a volatile substance in the unfermented apple pomace.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-methyl-2-furoate is an important raw material used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.It is a volatile substance in the unfermented apple pomace.

Solubility
Insoluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 6141-57-7
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14
MDL Number MFCD00014110
InChI Key AQQYRDKMXXSIMP-UHFFFAOYSA-N
Synonym methyl 3-methyl-2-furoate, methyl 3-methylfuroate, methyl 3-methyl-2-furancarboxylate, furancarboxylic acid, methyl-, methyl ester, 3-methyl-2-furancarboxylic acid methyl ester, 3-methyl-furan-2-carboxylic acid methyl ester, 2-furoic acid, 3-methyl-, methyl ester, acmc-20a9f6, methyl-3-methyl-2-furoate, 2-furancarboxylic acid, 3-methyl-, methyl ester
PubChem CID 350231
IUPAC Name methyl 3-methylfuran-2-carboxylate
SMILES COC(=O)C1=C(C)C=CO1

Specifications

Melting Point 33°C to 37°C
Boiling Point 60°C (1 mmHg)
Flash Point 82°C (180°F)
Quantity 25 g
Beilstein 116835
Solubility Information Insoluble in water.
Formula Weight 140.14
Percent Purity 98%
Chemical Name or Material Methyl 3-methyl-2-furoate
EINECSNumber 228-131-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only