missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Methyl-2-furancarboxylate 98.0+%, TCI America™

Catalog No. M23775G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23775G 5 g
M23771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23775G Supplier TCI America Supplier No. M23775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6141-57-7
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14
MDL Number MFCD00014110
InChI Key AQQYRDKMXXSIMP-UHFFFAOYSA-N
Synonym methyl 3-methyl-2-furoate, methyl 3-methylfuroate, methyl 3-methyl-2-furancarboxylate, furancarboxylic acid, methyl-, methyl ester, 3-methyl-2-furancarboxylic acid methyl ester, 3-methyl-furan-2-carboxylic acid methyl ester, 2-furoic acid, 3-methyl-, methyl ester, acmc-20a9f6, methyl-3-methyl-2-furoate, 2-furancarboxylic acid, 3-methyl-, methyl ester
PubChem CID 350231
IUPAC Name methyl 3-methylfuran-2-carboxylate
SMILES COC(=O)C1=C(C)C=CO1

Specifications

Melting Point 38°C
Color White-Yellow
Boiling Point 74°C
Quantity 5 g
Formula Weight 140.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Methyl-2-furancarboxylate