missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Methoxyacrylate 95.0+%, TCI America™

Catalog No. M0995500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0995500ML 500 mL
M099525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0995500ML Supplier TCI America Supplier No. M0995500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34846-90-7
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00128206
InChI Key AUTCCPQKLPMHDN-ONEGZZNKSA-N
Synonym methyl 3-methoxyacrylate, methyl trans-3-methoxyacrylate, methyl 2e-3-methoxyprop-2-enoate, e-methyl 3-methoxyacrylate, 3-methoxyacrylic acid methyl ester, methyl-3-methoxyacrylate, 2-propenoic acid, 3-methoxy-, methyl ester, 2e, methyl trans-3-methoxy acrylate, e-methyl-3-methoxyacrylate, methyl-trans-3-methoxyacrylate
PubChem CID 5323651
IUPAC Name methyl (E)-3-methoxyprop-2-enoate
SMILES COC=CC(=O)OC

Specifications

Color Yellow
Boiling Point 56°C
Quantity 500 mL
Formula Weight 116.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Methoxyacrylate
EINECSNumber (2)-3565
TSCA No