Learn More
Methyl 3-methoxy-2-nitrobenzoate, 98%, Thermo Scientific Chemicals
Description
Methyl 3-methoxy-2-nitrobenzoate is used as Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 3-methoxy-2-nitrobenzoate is used as Pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 142°C to 144°C |
| Quantity | 5 g |
| Beilstein | 2697098 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 211.18 |
| Percent Purity | 98% |
| Chemical Name or Material | Methyl 3-methoxy-2-nitrobenzoate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only