missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-Iodo-4-methylbenzoate 98.0+%, TCI America™
Supplier: TCI America M269425G
Specifications
| Methyl 3-Iodo-4-methylbenzoate | |
| 28°C | |
| C9H9IO2 | |
| 25 g | |
| NKMHAOTZPFVSPC-UHFFFAOYSA-N | |
| methyl 3-iodo-4-methylbenzoate | |
| 13614033 | |
| ≥98.0% (GC) |
| 90347-66-3 | |
| White-Yellow | |
| MFCD00230583 | |
| 3-iodo-4-methylbenzoic acid methyl ester, methyl 3-iodo-p-toluate, methyl-3-iodo-4-methylbenzoate, methyl 3-iodo-4-methyl-benzoate, 2-iodo-4-methoxycarbonyl toluene, benzoic acid, 3-iodo-4-methyl-, methyl ester, methyl3-iodo-4-methylbenzoate, 3-iodo-4-methylbenzoic acidmethyl ester, methyl 3-iodo-4-meth, ksc495m1r | |
| CC1=C(C=C(C=C1)C(=O)OC)I | |
| 276.073 | |
| 276.07 | |
| Crystalline Lumps |
Chemical Identifiers
| 90347-66-3 | |
| 276.073 | |
| NKMHAOTZPFVSPC-UHFFFAOYSA-N | |
| 13614033 | |
| CC1=C(C=C(C=C1)C(=O)OC)I |
| C9H9IO2 | |
| MFCD00230583 | |
| 3-iodo-4-methylbenzoic acid methyl ester, methyl 3-iodo-p-toluate, methyl-3-iodo-4-methylbenzoate, methyl 3-iodo-4-methyl-benzoate, 2-iodo-4-methoxycarbonyl toluene, benzoic acid, 3-iodo-4-methyl-, methyl ester, methyl3-iodo-4-methylbenzoate, 3-iodo-4-methylbenzoic acidmethyl ester, methyl 3-iodo-4-meth, ksc495m1r | |
| methyl 3-iodo-4-methylbenzoate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator