accessibility menu, dialog, popup

UserName

CH3OOCH3I

CAS RN 90347-66-3

IUPAC Name: methyl 3-iodo-4-methylbenzoate
Synonyms: methyl 3-iodo-4-methylbenzoate
Molecular Weight (g/mol): 276.07
Molecular Formula: C9H9IO2
InChi Key: NKMHAOTZPFVSPC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(I)=C(C)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)