missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Iodo-4-methylbenzoate 98.0+%, TCI America™

Catalog No. p-7133374
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26945G 5 g
M269425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26945G Supplier TCI America Supplier No. M26945G
Only null left

Chemical Identifiers

CAS 90347-66-3
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.073
MDL Number MFCD00230583
InChI Key NKMHAOTZPFVSPC-UHFFFAOYSA-N
Synonym 3-iodo-4-methylbenzoic acid methyl ester, methyl 3-iodo-p-toluate, methyl-3-iodo-4-methylbenzoate, methyl 3-iodo-4-methyl-benzoate, 2-iodo-4-methoxycarbonyl toluene, benzoic acid, 3-iodo-4-methyl-, methyl ester, methyl3-iodo-4-methylbenzoate, 3-iodo-4-methylbenzoic acidmethyl ester, methyl 3-iodo-4-meth, ksc495m1r
PubChem CID 13614033
IUPAC Name methyl 3-iodo-4-methylbenzoate
SMILES CC1=C(C=C(C=C1)C(=O)OC)I

Specifications

Melting Point 28°C
Color White-Yellow
Quantity 5 g
Formula Weight 276.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 3-Iodo-4-methylbenzoate
TSCA No
Recommended Storage Refrigerator