missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Iodo-4-methoxybenzoate 98.0+%, TCI America™

Catalog No. M20965G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20965G Supplier TCI America Supplier No. M20965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35387-93-0
Molecular Formula C9H9IO3
Molecular Weight (g/mol) 292.07
MDL Number MFCD00052925
InChI Key GHNGBFHLUOJHKP-UHFFFAOYSA-N
Synonym 3-iodo-4-methoxy-benzoic acid methyl ester, 3-iodo-4-methoxybenzoic acid methyl ester, benzoic acid, 3-iodo-4-methoxy-, methyl ester, methyl3-iodo-4-methoxybenzoate, methyl 3-iodo-p-anisate, acmc-1agb4, ksc495q3h, methyl-3-iodo-4-methoxybenzoate, methyl 3-iodo-4-methoxy-benzoate, 3-iodo-p-anisic acid methyl ester
PubChem CID 316980
IUPAC Name methyl 3-iodo-4-methoxybenzoate
SMILES COC(=O)C1=CC=C(OC)C(I)=C1

Specifications

Melting Point 95°C
Quantity 5 g
Formula Weight 292.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Iodo-4-methoxybenzoate