Learn More
Methyl 3-hydroxybenzoate, 99%
CAS: 19438-10-9 | C8H8O3 | 152.15 g/mol
Supplier: Thermo Scientific Chemicals A1442536
Description
Methyl 3-hydroxybenzoate is used in the preparation of 3-hydroxybenzene-1,2-dicarbaldehyde and O-methyl O-[3-methyl-4-(methylthio)phenyl]O-(3-methylcarboxyphenyl) phosphorothioate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 3-hydroxybenzoate is used in the preparation of 3-hydroxybenzene-1,2-dicarbaldehyde and O-methyl O-[3-methyl-4-(methylthio)phenyl]O-(3-methylcarboxyphenyl) phosphorothioate.
Solubility
Soluble in chloroform, dimetyl Sulfoxide,ethanol and methanol.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Methyl 3-hydroxybenzoate | |
| 70°C to 72°C | |
| C8H8O3 | |
| 500 g | |
| methyl m-hydroxybenzoate, 3-hydroxybenzoic acid methyl ester, m-carbomethoxyphenol, methyl m-oxybenzoate, m-hydroxybenzoic acid methyl ester, benzoic acid, 3-hydroxy-, methyl ester, 3-hydroxy-benzoic acid methyl ester, unii-569m9g3stj, 3-methoxycarbonyl phenol, benzoic acid, m-hydroxy-, methyl ester | |
| YKUCHDXIBAQWSF-UHFFFAOYSA-N | |
| methyl 3-hydroxybenzoate | |
| 88068 | |
| 99% |
| 19438-10-9 | |
| 280°C to 281°C (708 mmHg) | |
| MFCD00002295 | |
| 2208129 | |
| Soluble in chloroform,dimetyl Sulfoxide,ethanol and methanol. | |
| COC(=O)C1=CC=CC(O)=C1 | |
| 152.15 | |
| 152.15 |
Chemical Identifiers
| 19438-10-9 | |
| 152.15 | |
| YKUCHDXIBAQWSF-UHFFFAOYSA-N | |
| 88068 | |
| COC(=O)C1=CC=CC(O)=C1 |
| C8H8O3 | |
| MFCD00002295 | |
| methyl m-hydroxybenzoate, 3-hydroxybenzoic acid methyl ester, m-carbomethoxyphenol, methyl m-oxybenzoate, m-hydroxybenzoic acid methyl ester, benzoic acid, 3-hydroxy-, methyl ester, 3-hydroxy-benzoic acid methyl ester, unii-569m9g3stj, 3-methoxycarbonyl phenol, benzoic acid, m-hydroxy-, methyl ester | |
| methyl 3-hydroxybenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 243-071-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only