missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hydroxybenzoate, 99%

Catalog No. p-7021900
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1442514 25 g
AAA1442522 100 g
AAA1442536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1442514 Supplier Thermo Scientific Chemicals Supplier No. A1442514

CAS: 19438-10-9 | C8H8O3 | 152.15 g/mol

Methyl 3-hydroxybenzoate is used in the preparation of 3-hydroxybenzene-1,2-dicarbaldehyde and O-methyl O-[3-methyl-4-(methylthio)phenyl]O-(3-methylcarboxyphenyl) phosphorothioate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-hydroxybenzoate is used in the preparation of 3-hydroxybenzene-1,2-dicarbaldehyde and O-methyl O-[3-methyl-4-(methylthio)phenyl]O-(3-methylcarboxyphenyl) phosphorothioate.

Solubility
Soluble in chloroform, dimetyl Sulfoxide,ethanol and methanol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 19438-10-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002295
InChI Key YKUCHDXIBAQWSF-UHFFFAOYSA-N
Synonym methyl m-hydroxybenzoate, 3-hydroxybenzoic acid methyl ester, m-carbomethoxyphenol, methyl m-oxybenzoate, m-hydroxybenzoic acid methyl ester, benzoic acid, 3-hydroxy-, methyl ester, 3-hydroxy-benzoic acid methyl ester, unii-569m9g3stj, 3-methoxycarbonyl phenol, benzoic acid, m-hydroxy-, methyl ester
PubChem CID 88068
IUPAC Name methyl 3-hydroxybenzoate
SMILES COC(=O)C1=CC=CC(O)=C1

Specifications

Melting Point 70°C to 72°C
Boiling Point 280°C to 281°C (708 mmHg)
Quantity 25 g
Beilstein 2208129
Solubility Information Soluble in chloroform,dimetyl Sulfoxide,ethanol and methanol.
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material Methyl 3-hydroxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 243-071-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only