missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-cyano-4-methoxybenzoate, 97%

Catalog No. AAL1110706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1110706 5 g
AAL1110703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1110706 Supplier Thermo Scientific Chemicals Supplier No. L1110706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25978-74-9 | C10H9NO3 | 191.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25978-74-9
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00052930
InChI Key RYJSFYBJYKFNCF-UHFFFAOYSA-N
Synonym benzoic acid, 3-cyano-4-methoxy-, methyl ester, acmc-1cj4t, methyl 3-cyano-4-methoxy-benzoate, methyl 3-cyano-4-methoxybenzoate., methyl 3-cyano-4-methyloxy benzoate, 3-cyano-4-methoxy-benzoic acid methyl ester, benzoicacid,3-cyano-4-methoxy-,methyl ester
PubChem CID 2801011
IUPAC Name methyl 3-cyano-4-methoxybenzoate
SMILES COC1=C(C=C(C=C1)C(=O)OC)C#N

Specifications

Melting Point 123°C to 127°C
Quantity 5 g
UN Number UN3439
Beilstein 2374769
Formula Weight 191.19
Percent Purity 97%
Chemical Name or Material Methyl 3-cyano-4-methoxybenzoate
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only