missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-chlorobenzoate, 98%

Catalog No. AAB2137122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2137122 100 g
AAB2137114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2137122 Supplier Thermo Scientific Chemicals Supplier No. B2137122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2905-65-9 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2905-65-9
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00000592
InChI Key XRDRKVPNHIWTBX-UHFFFAOYSA-N
Synonym methyl m-chlorobenzoate, benzoic acid, 3-chloro-, methyl ester, m-chlorobenzoic acid methyl ester, methyl-3-chlorobenzoate, benzoic acid, m-chloro-, methyl ester, 3-chlorobenzoic acid methyl ester, 3-chloro-benzoic acid methyl ester, acmc-1cegg, dsstox_cid_7507, 3-chlorobenzoic acid methyl
PubChem CID 17946
IUPAC Name methyl 3-chlorobenzoate
SMILES COC(=O)C1=CC=CC(Cl)=C1

Specifications

Melting Point 20°C to 22°C
Density 1.227
Boiling Point 99°C to 101°C (12 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.531
Quantity 100 g
Beilstein 2045126
Merck Index 14,2123
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material Methyl 3-chlorobenzoate
Show More Show Less
Hazard Category H302-H315-H319
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 220-814-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only